missing translation for 'onlineSavingsMsg'
Learn More

Slice™ Self-Opening Scissors

Product Code. 16483649
Change view
Click to view available options
Color:
Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16483649 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16483649 Supplier Slice™ Supplier No. 10595
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for industrial tailors and fibre artists, or anyone with arthritis, hand weakness, or dexterity challenges

  • Zirconium oxide Slice™ safety blades ideal for thread and fibres
  • Durable reinforced nylon handles with ribbed grips
  • Safer than traditional scissors
  • Self-open design lowers muscle effort
  • Finger-friendly™ edge lasts 11x longer than metal
  • Ambidextrous design
  • Reduces injuries, lowers costs
  • Non-sparking, non-conductive, non-magnetic blades
  • Chemically inert blades never rust
  • Oil, lubricant, and maintenance free
  • Easy to clean
  • BPA, phthalate, and lead-free scissors
  • LIghtweight and compact
  • Features lanyard hole and safety cap

Specifications

Product Type Scissors
Material PA6 GF50, Polypropylene,Stainless Steel, Zirconium Oxide
Color Gray
For Use With (Application) Safety Cutting
Includes Ceramic blades
No. per Pack 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.