missing translation for 'onlineSavingsMsg'
Learn More

Selexipag, MedChemExpress

Product Code. 16458817
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458817 5 mg 5mg
16416228 10 mg 10mg
16402357 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16458817 Supplier Medchem Express Supplier No. HY14870
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Selexipag (NS-304) is an orally available and potent agonist for the Prostacyclin (PGI2) receptor (IP receptor).

Selexipag (NS-304) is an orally available and potent agonist for the Prostacyclin (PGI2) receptor (IP receptor).

Specifications

Chemical Name or Material Selexipag
CAS 475086-01-2
Color White
Purity Grade Notes Research
Molecular Formula C26H32N4O4S
Quantity 5 mg
Synonym NS-304 ACT-293987
Solubility Information DMSO : ≥ 50 mg/mL (100.68 mM)
SMILES O=C(NS(=O)(C)=O)COCCCCN(C1=NC(C2=CC=CC=C2)=C(C3=CC=CC=C3)N=C1)C(C)C
Molecular Weight (g/mol) 496.62
Formula Weight 496.62
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.