missing translation for 'onlineSavingsMsg'
Learn More

Selenous acid, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11347007
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
7783-00-8
H2O3Se
128.98
MFCD00011331
MCAHWIHFGHIESP-UHFFFAOYSA-N
selenious acid, selenious acid h2seo3, monohydrated selenium dioxide, unii-f6a27p4q4r, selenium dioxide, monohydrated, ccris 5530, selenious acid usp, selenious acid,selenous acid,monohydrated selenium dioxide, selenium, reference standard solution
1091
CHEBI:26642
selenous acid
O[Se](O)=O
This item is not returnable. View return policy
7783-00-8
H2O3Se
128.98
MFCD00011331
MCAHWIHFGHIESP-UHFFFAOYSA-N
selenious acid, selenious acid h2seo3, monohydrated selenium dioxide, unii-f6a27p4q4r, selenium dioxide, monohydrated, ccris 5530, selenious acid usp, selenious acid,selenous acid,monohydrated selenium dioxide, selenium, reference standard solution
1091
CHEBI:26642
selenous acid
O[Se](O)=O

Selenous acid is used in steel guns, wherein it changes the color of the steel from silver-grey to blue-grey. It serves as an important component of the Mecke reagent, which is utilized for drug testing. It acts as a source of selenium. It acts as a reagent for alkaloids and an oxidizing agent. Further, it is used in labeling radiopharmaceuticals. It is involved in the organic synthesis for the preparation of glyoxal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 70°C (decomposition)
Odor Odorless
Sensitivity Hygroscopic
Quantity 25 g
Physical Form Crystalline Aggregates
UN Number UN3290
Merck Index 14,8430
Solubility Information Soluble in water and ethanol.
Formula Weight 128.97
Percent Purity 99.999%
Grade Puratronic™
Assay (metals basis)
Chemical Name or Material Selenous acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'