missing translation for 'onlineSavingsMsg'
Learn More

Selenourea, TRC

Product Code. 30137610
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137610 5 g 5g
30003597 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137610 Supplier Toronto Research Chemicals Supplier No. TRCS2531035G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-10-4
Molecular Formula CH4N2Se
Molecular Weight (g/mol) 123.02
Synonym 2-Selenourea, Carbamimidoselenoic Acid, Isoselenourea, NSC 227840, Selenouronium;
IUPAC Name selenourea
SMILES NC(=[Se])N

Specifications

Quantity 5 g
InChI Formula InChI=1S/CH4N2Se/c2-1(3)4/h(H4,2,3,4)
Formula Weight 123.954
Chemical Name or Material Selenourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.