missing translation for 'onlineSavingsMsg'
Learn More

Selenourea, 99%

Product Code. 11384247
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384247 1 g 1g
11394247 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384247 Supplier Thermo Scientific Alfa Aesar Supplier No. 036720.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 630-10-4 | CH3N2Se | 122.02 g/mol

Selenourea is used in the synthesis of selenium heterocycles, which exhibit anti-inflammatory and anti-tumor activity. It acts as an effective ligand and form complexes with transition metals and metalloids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-10-4
Molecular Formula CH3N2Se
Molecular Weight (g/mol) 122.02
MDL Number MFCD00008065
InChI Key ZUCFIOXGLBMWHV-UHFFFAOYSA-N
Synonym selenourea, 2-selenourea, carbamimidoselenoic acid, selenouronium, urea, seleno, rcra waste number p103, rcra waste no. p103, iykvlicpfcezof-uhfffaoysa-n, ch4n2se, isoselenourea
PubChem CID 6327594
IUPAC Name $l^{1}-selanylmethanimidamide
SMILES C(=N)(N)[Se]

Specifications

Melting Point ∼210°C (decomposition)
Color White to Gray
Packaging Ampule
Quantity 1 g
UN Number UN3283
Sensitivity Air and moisture sensitive
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 123.02
Percent Purity 99.97%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Selenourea
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.