missing translation for 'onlineSavingsMsg'
Learn More

Selenophene, 99%

Product Code. 10269543 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10269543 10 g Glass bottle 10g
10581301 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10269543 Supplier Thermo Scientific Acros Supplier No. 271900100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 288-05-1 | C4H4Se | 131.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-05-1
Molecular Formula C4H4Se
Molecular Weight (g/mol) 131.04
MDL Number MFCD00075487
InChI Key MABNMNVCOAICNO-UHFFFAOYSA-N
Synonym selenophen, selenophene, acmc-1cr2h
PubChem CID 136130
ChEBI CHEBI:30857
IUPAC Name selenophene
SMILES C1=C[Se]C=C1

Specifications

Melting Point -38.0°C
Color Colorless to Yellow
Density 1.5200g/mL
Boiling Point 109.9°C to 110.1°C (752.0 mmHg)
Flash Point −2°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5790 to 1.5810
Quantity 10 g
UN Number 1992
Specific Gravity 1.52
Solubility Information Solubility in water: .. Other solubilities: azeotrope with water, soluble in all organic solvents
Formula Weight 131.04
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Selenophene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.