missing translation for 'onlineSavingsMsg'
Learn More

Selenomethionine, MedChemExpress

Product Code. p-200097619
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452236 500 mg 500mg
16418556 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452236 Supplier Medchem Express Supplier No. HYB1000
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Selenomethionine is a naturally occurring amino acid containing selenium, is a common natural food source of selenium.

Selenomethionine is a naturally occurring amino acid containing selenium, is a common natural food source of selenium.

Specifications

Chemical Name or Material Selenomethionine
CAS 1464-42-2
Color White
Purity Grade Notes Research
Molecular Formula C5H11NO2Se
Synonym Seleno-DL-methionine DL-Selenomethionine
Solubility Information H2O : 12.5 mg/mL (63.74 mM; Need ultrasonic)
SMILES O=C(O)C(N)CC[Se]C
Molecular Weight (g/mol) 196.11
Quantity 500 mg
Formula Weight 196.11
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.