missing translation for 'onlineSavingsMsg'
Learn More

Selenium sulfide, 94%

Product Code. 11380208
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11380208

Brand: Thermo Scientific Alfa Aesar 042943.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7488-56-4 | S2Se | 143.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7488-56-4
Molecular Formula S2Se
Molecular Weight (g/mol) 143.09
MDL Number MFCD00011227
InChI Key JNMWHTHYDQTDQZ-UHFFFAOYSA-N
Synonym selenium disulfide, selenium disulphide, selsun blue, selenium iv disulfide, selenium sulfide ses2, sebusan, selenex, selenix, selukos, lenium
PubChem CID 24087
IUPAC Name thione
SMILES S=[Se]=S

Specifications

Boiling Point (decomposition)
Melting Point <100°C
Color Red to Brown
Physical Form Powder
Quantity 100 g
Assay Percent Range 94%
Solubility Information Insoluble in water,in dilute HCl
Formula Weight 143.09
Chemical Name or Material Selenium sulfide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.