missing translation for 'onlineSavingsMsg'
Learn More

Selenium Standard: Se @ 1000 μg/mL in 5% HNO3, VHG™

Product Code. 17937109
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17937109 100 mL 100mL
17938309 50 mL 50mL
17979309 250 mL 250mL
17957109 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17937109 Supplier VHG Labs Supplier No. VHGPSEN100

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Selenium Standard: Se @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Selenium
CAS 7782-49-2
Methods ICP/ICP-MS
Analyte or Component Names Selenium
Shipping Condition Room Temperature
Molecular Formula Se
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym elenium, elemental, Vandex
InChI Formula InChI=1S/Se
SMILES [Se]
IUPAC Name selenium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 78.96
Quantity 100 mL
Formula Weight 79.9165 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.