missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selenium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
£52.65
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10458600
|
Fisher Chemical
J/8061/08 |
500 mL | |||||||
Description
Chemical Identifiers
134090 |
Specifications
HDPE plastic bottle | |
134090 | |
Liquid |
Colorless | |
1 |