missing translation for 'onlineSavingsMsg'
Learn More

Euro Protection™ Selenium Safety Boots

Product Code. 12054535
Click to view available options
:
Size: 46
This item is not returnable. View return policy

Product Code. 12054535

Brand: Euro Protection™ 9SELE46

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Euro Protection™ Selenium Safety Boots are economical, easy to wear, and constructed of resistant PVC/nitrile.

  • Suitable for agriculture/farming industry
  • Easy-on and easy-off
  • Good price to quality ratio
  • PVC upper
  • Polyester lining
  • PVC / Nitrile outsole

Specifications

Color Green, Black
For Use With (Application) Agriculture/farming
Liner Material Polyester
Material PVC
Size 46
Sole Material PVC, Nitrile
Weight (Metric) 1165 g per boot in size 9.5
Product Type Safety Boot
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.