missing translation for 'onlineSavingsMsg'
Learn More

Selenium(IV) oxychloride, 97%, Thermo Scientific™

Product Code. 10577943
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 180.0°C
Melting Point 9.0°C
CAS 7791-23-3
Color Yellow
Physical Form Liquid
Quantity 5 g
Assay Percent Range 97%
Molecular Formula Cl2OSe
Linear Formula SeOCl2
MDL Number MFCD00016327
Synonym selenium oxychloride, seleninyl chloride, selenium oxydichloride, seleninyl dichloride, selenium chloride oxide, hsdb 678, selenium chloride oxide secl2o, selenium iv oxychloride, selenium dichloride oxide, selenoxychlorid
Solubility Information Solubility in water: reacts.
InChI Key LIRMPIXWGWLNND-UHFFFAOYSA-N
SMILES O=[Se](Cl)Cl
Molecular Weight (g/mol) 165.87
PubChem CID 24647
Formula Weight 165.86
Percent Purity 97%
Packaging Glass bottle
Specific Gravity 2.43
Density 2.4300g/mL
Chemical Name or Material Selenium(IV) oxychloride, 97%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'