missing translation for 'onlineSavingsMsg'
Learn More

Selenium(IV) oxide, Puratronic™, 99.999% (metals basis)

Product Code. 11317007
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317007 25 g 25g
11327007 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317007 Supplier Thermo Scientific Alfa Aesar Supplier No. 010854.14
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 7446-08-4 | O2Se | 110.97 g/mol

In organic reactions, selenium dioxide is often used as an oxidizing reagent employed for carrying out allylic oxidations, Riley oxidation, synthesis of diones, etc. It is used to impart a red color to glass. It finds use as a toner in photographic developing, and an active ingredient in some cold-blueing solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-08-4
Molecular Formula O2Se
Molecular Weight (g/mol) 110.97
MDL Number MFCD00003562
InChI Key JPJALAQPGMAKDF-UHFFFAOYSA-N
Synonym selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
PubChem CID 24007
IUPAC Name selenium dioxide
SMILES O=[Se]=O

Specifications

Melting Point 340°C to 350°C (sublimation)
Physical Form Lump/Powder
Quantity 25 g
UN Number UN3283
Merck Index 14,8434
Solubility Information Soluble in water,benzene,ethanol and methanol. Sprangly soluble in acetone and acetic acid.
Formula Weight 110.96
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Assay (metals basis)
Chemical Name or Material Selenium(IV) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.