missing translation for 'onlineSavingsMsg'
Learn More

Selenium(IV) chloride, 99.5% (metals basis)

Product Code. p-7049166
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11342188

Brand: Thermo Scientific Alfa Aesar 013096.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10026-03-6 | Cl4Se | 220.771 g/mol

It is used in the synthesis of other selenium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-03-6
Molecular Formula Cl4Se
Molecular Weight (g/mol) 220.771
MDL Number MFCD00011225
InChI Key LNBXMNQCXXEHFT-UHFFFAOYSA-N
Synonym selenium tetrachloride, selenium chloride secl4 , t-4, unii-4gb8868p5j, selenium iv chloride 1:4, secl4, tetrachloro-, e4-selane, selenium iv chloride, selenium iv chloride-8 mesh, selenium iv chloride trace metals basis 25g
PubChem CID 66205
SMILES Cl[Se](Cl)(Cl)Cl

Specifications

Melting Point (sublimation)
Physical Form Lump
Quantity 25 g
UN Number UN2923
Merck Index 14,8438
Solubility Information Decomposes in water and moist air; Insoluble in liquid bromine; Sublimes when heated
Formula Weight 220.77
Percent Purity 99.5%
Odor Pungent
Refractive Index 1.807
Sensitivity Moisture sensitive
Density 2.600 g/mL
Assay (metals basis)
Chemical Name or Material Selenium(IV) chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.