missing translation for 'onlineSavingsMsg'
Learn More

Selenium(IV) bromide, 99+%

Product Code. 11333758
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333758 25 g 25g
11470251 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333758 Supplier Thermo Scientific Alfa Aesar Supplier No. 040240.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-65-3 | Br4Se | 398.587 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-65-3
Molecular Formula Br4Se
Molecular Weight (g/mol) 398.587
MDL Number MFCD00049766
InChI Key VTQZBGAODFEJOW-UHFFFAOYSA-N
Synonym selenium tetrabromide, selenium iv bromide, selenium bromide sebr4 , t-4, unii-8o54yxr180, tetrabromo-, e4-selane, sebr4, tetrabromo-$l^ 4-selane, tetrabromo-??-selane, selenium iv bromide 5g
PubChem CID 82246
SMILES [Se](Br)(Br)(Br)Br

Specifications

Melting Point 70°C to 80°C (decomposition)
Physical Form Powder
Quantity 25 g
UN Number UN2923
Merck Index 14,8437
Solubility Information Decomposes in water; Soluble in carbon disulfide, chloroform, ethyl bromide
Formula Weight 398.6
Percent Purity ≥99%
Sensitivity Moisture sensitive
Chemical Name or Material Selenium(IV) bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.