missing translation for 'onlineSavingsMsg'
Learn More

Selenium Isotopic Standard: Se @ 10 μg/mL in 2% HNO3 (Enriched in Se-78), VHG™

Product Code. 17913376
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Product Code. Quantity unitSize
17913376 50 mL 50mL
1 options
This item is not returnable. View return policy

Product Code. 17913376

Brand: VHG Labs VHGLIS78SE50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Selenium Isotopic Standard: Se @ 10 μg/mL in 2% HNO3 (Enriched in Se-78)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Selenium 78
CAS 14833-16-0
Methods ICP-MS
Analyte or Component Names Selenium 78
Shipping Condition Room Temperature
Molecular Formula 78Se
Synonym 78Se
SMILES [78Se]
IUPAC Name Selenium 78
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 77.92
Quantity 50 mL
Formula Weight 77.9173 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.