missing translation for 'onlineSavingsMsg'
Learn More

Selenium dichloride oxide, 99% (metals basis)

Product Code. 11344789
Change view
Click to view available options
Quantity:
10 mL
25 mL
Unit Size:
10mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344789 25 mL 25mL
11354789 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344789 Supplier Thermo Scientific Alfa Aesar Supplier No. 068108.AC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7791-23-3 | Cl2OSe | 165.87 g/mol

Selenium(IV) chloride oxide is used as a polar solvent for synthetic phenolic resins. It is also used to prepare selenium oxybromide and selenium nitride. It acts as a chlorinating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-23-3
Molecular Formula Cl2OSe
Molecular Weight (g/mol) 165.87
MDL Number MFCD00016327
InChI Key LIRMPIXWGWLNND-UHFFFAOYSA-N
Synonym selenium oxychloride, seleninyl chloride, selenium oxydichloride, seleninyl dichloride, selenium chloride oxide, hsdb 678, selenium chloride oxide secl2o, selenium iv oxychloride, selenium dichloride oxide, selenoxychlorid
PubChem CID 24647
SMILES O=[Se](Cl)Cl

Specifications

Boiling Point 176.4°C
Melting Point 8.5°C
Color Yellow
Physical Form Liquid
Quantity 25 mL
UN Number UN2879
Merck Index 14,8435
Solubility Information Miscible with chloroform,carbon disulphide and benzene.
Formula Weight 165.87
Percent Purity 99%
Refractive Index 1.651
Sensitivity Moisture sensitive
Density 2.42 g/mL
Assay (metals basis)
Chemical Name or Material Selenium dichloride oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.