missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Selenite Broth Base (Lactose) (Dehydrated)

Product Code. 10219632
Click to view available options
Form:
Powder
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10219632

Brand: Thermo Scientific™ CM0395B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enrich and isolate Salmonella from food and feces with this medium.

Enrich and isolate Salmonella from feces and food products with Thermo Scientific™ Oxoid™ Selenite Broth Base (Lactose).The presence of Selenium in the medium provides selectivity against non-target bacteria.

Selenite Broth Base contains lactose as a source of fermentable carbohydrate which helps to prevent any rise in pH value during incubation, thereby preventing any loss of selective activity of selenite.

Use Selenite Broth Base for enrichment and isolation of Salmonella from feces, urine, water, foods, and materials of sanitary importance.

  • Selective against non-target Gram-positive flora in the first few hours of incubation
  • Routinely employed for detection of Salmonella from fecal specimens since Salmonella usually represent only a small percentage of the intestinal flora present

Klett1 first demonstrated the selective inhibitory effects of selenite, and Guth2 used it to isolate Salmonella typhi. It was 20 years later before Leifson3 fully investigated selenite and promoted wide use of the medium.

Proteus and Pseudomonas species appear to be resistant to its effects4. Lactose is added as a fermentable carbohydrate to prevent a rise in pH value during incubation because any increase in pH will reduce the selective activity of selenite. The fact that Proteus and Pseudomonas species do not ferment lactose may explain why they escape inhibition.

To minimise any possible risk of teratogenicity to laboratory workers, the sodium biselenite must be added as a solution to this medium. Robertson5 reported miscarriages and possible teratogenic effects on pregnant laboratory assistants which may have been caused by ingested sodium biselenite. This substance has, therefore, been removed from the Oxoid powdered medium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 30°C
Description Selenite Broth Base (Lactose) (Dehydrated)
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.