missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELENBP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27230-1-AP-20UL
This item is not returnable.
View return policy
Description
Selenium is an essential trace element that confers tolerance to toxicity arising through exposure to heavy metals or other reactive xenobiotics. Selenium exhibits potent anticarcinogenic properties, and deficiency of selenium may cause certain neurologic diseases. Both effects are attributed to selenium-binding proteins. Selenium binding protein 1 is down-regulated in lung adenocarcinoma, colorectal cander and ovarian cancer. It is two-fold upregulated in the brains of patients suffering from schizophrenia, and is therefore a biomarker for this disease.Specifications
| SELENBP1 | |
| Polyclonal | |
| Unconjugated | |
| SELENBP1 | |
| hSBP, hSP56, LPSB, SBP, SBP56, SELENBP1, selenium binding protein 1, SP56 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8991 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13228 | |
| SELENBP1 | |
| SELENBP1 Fusion Protein Ag20967 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction