missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SELENBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55263
This item is not returnable.
View return policy
Description
SELENBP1 Polyclonal specifically detects SELENBP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SELENBP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| 56 kDa selenium-binding protein, hSBP, hSP56, LPSB, SBP, SBP56, selenium binding protein 1, selenium-binding protein 1, SP56FLJ13813 | |
| Rabbit | |
| 44 kDa | |
| 100 μL | |
| Primary | |
| Recognizes C-terminal region on both isoforms of SELENBP1. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Q13228 | |
| SELENBP1 | |
| Synthetic peptides corresponding to SELENBP1(selenium binding protein 1) The peptide sequence was selected from the C terminal of SELENBP1 (AAH32997). Peptide sequence KQFYPDLIREGSVMLQVDVDTVKGGLKLNPNFLVDFGKEPLGPALAHELR. | |
| Affinity purified | |
| RUO | |
| 8991 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction