missing translation for 'onlineSavingsMsg'
Learn More

Selegiline Hydrochloride, LoGiCal

Product Code. 30457798
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
missing translation for 'unitSize':
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30457798 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30457798 missing translation for 'mfr' LoGiCal missing translation for 'supplierNo' LGCFOR0216.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Selegiline Hydrochloride
CAS 14611-52-0
Analyte or Component Names Selegiline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C13 H17 N . Cl H
Certifications/Compliance ISO 9001
Synonym Selegiline Hydrochloride, (-)-Deprenil hydrochloride, (-)-Deprenyl hydrochloride, Eldeprine, Eldepryl, FP Tablet 2.5, Jumex, Movergan, Plurimen, R-Selegiline hydrochloride, Tonus, l-Deprenyl hydrochloride, Benzeneethanamine, N,alpha-dimethyl-N-2-propyn-1-yl-, hydrochloride (1:1), (alphaR)-, Benzeneethanamine, N,alpha-dimethyl-N-2-propynyl-, hydrochloride, (R)-, Benzeneethanamine, N,alpha-dimethyl-N-2-propynyl-, hydrochloride, (alphaR)- (9CI)
InChI Formula InChI=1S/C13H17N.ClH/c1-4-10-14(3)12(2)11-13-8-6-5-7-9-13;/h1,5-9,12H,10-11H2,2-3H3;1H/t12-;/m1./s1
Solution Type Neat
SMILES Cl.C[C@H](Cc1ccccc1)N(C)CC#C
IUPAC Name (2R)-N-methyl-1-phenyl-N-prop-2-ynylpropan-2-amine;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 223.74
Quantity 10 mg
Formula Weight 223.1128 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.