missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEL1L3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144785
This item is not returnable.
View return policy
Description
Immunogen sequence: ERCAEVQEIV SVYASAAKHG GERQEACHLH NSYLDLQRRY GRPSMCRAFP WEKELKDKHP SLFQALLEMD LLTVPRNQNE SVSEIGGKIF EKAVKRLSSI DGL Antigen sequence identity: Mouse 70%; Rat 67%.
SEL1L3 is a protein coding gene.
Specifications
| SEL1L3 | |
| Polyclonal | |
| Unconjugated | |
| SEL1L3 | |
| KIAA0746; Protein sel-1 homolog 3; sel-1 suppressor of lin-12-like 3; SEL1L family member 3; SEL1L3; Sel-1L3; Suppressor of lin-12-like protein 3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23231 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q68CR1 | |
| SEL1L3 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 449-551. Recombinant protein control fragment (Product #RP-106891). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction