missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEL1L Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-93463
This item is not returnable.
View return policy
Description
SEL1L Polyclonal specifically detects SEL1L in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| SEL1L | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| IBD2, PRO1063, protein sel-1 homolog 1, sel-1 (suppressor of lin-12, C.elegans)-like, sel-1 suppressor of lin-12-like (C. elegans), sel-1 suppressor of lin-12-like 1, Sel-1L, SEL1L1, SEL1-LIKE, Suppressor of lin 12 (sel-1), C. elegans, homolog of, Suppressor of lin-12-like protein 1, TSA305 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6400 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:1000 | |
| NP_005056.3 | |
| SEL1L | |
| Peptide with sequence C-TAYNSYKDGDYN corresponding to internal region according to NP_005056.3. | |
| 0.1 mg | |
| Primary | |
| Expected from sequence similarity: Dog, Cow and Pig. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction