missing translation for 'onlineSavingsMsg'
Learn More

SEL120-34A monohydrochloride, MedChemExpress

Product Code. 16465326
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16465326

Brand: Medchem Express HY111388A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SEL120-34A monohydrochloride is an ATP-competitive and selective CDK8 inhibitor, inhibits kinase activities of CDK8/CycC and CDK19/CycC complexes with IC50s of 4.4 nM and 10.4 nM, respectively, with a Kd of 3 nM for CDK8.

SEL120-34A monohydrochloride weakly inhibits CDK9 (calculated IC50=1070 nM), but shows no obvious activity against CDK1, 2, 4, 6, 5, 7. SEL120-34A monohydrochloride inhibits phosphorylation of STAT1 S727 and STAT5 S726. Has anti-tumor activity.

Specifications

Chemical Name or Material SEL120-34A monohydrochloride
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H19Br2ClN4
Quantity 100 mg
Solubility Information H2O : 50 mg/mL (110.96 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (37.00 mM; ultrasonic and warming and heat to 60°C)
SMILES CC1=C2C3=C(C(Br)=C1Br)CCCN3C(N4CCNCC4)=N2.Cl
Molecular Weight (g/mol) 450.6
Formula Weight 450.6
Percent Purity 98.51%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.