missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEL120-34A HCl, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
SEL120-34A HCl is a potent, selective, orally available, ATP-competitive CDK8 inhibitor, with IC50s of 4.4 nM and 10.4 nM for CDK8/CycC and CDK19/CycC, respectively, with antitumor activity.
Specifications
Specifications
| Chemical Name or Material | SEL120-34A HCl |
| CAS | 1609452-30-3 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C15H18Br2N4.xHCl |
| Quantity | 5 mg |
| Solubility Information | DMSO : 62.5 mg/mL (Need ultrasonic) |
| SMILES | CC1=C(Br)C(Br)=C2C3=C1N=C(N4CCNCC4)N3CCC2.[x HCl] |
| Percent Purity | 99.8% |
| Grade | Research |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction