missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Seipin Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547922
This item is not returnable.
View return policy
Description
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
This gene encodes protein seipin, which is located in the endoplasmic reticulum and may be important for lipid droplet morphology. Mutations in this gene have been associated with congenital generalized lipodystrophy type 2 or Berardinelli-Seip syndrome, a rare autosomal recessive disease characterized by a near absence of adipose tissue and severe insulin resistance. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| Seipin | |
| Polyclonal | |
| Unconjugated | |
| BSCL2 | |
| 2900097C17Rik; AI046355; Berardinelli-Seip congenital lipodystrophy 2 (seipin); Berardinelli-Seip congenital lipodystrophy 2 homolog (seipin); Bernardinelli-Seip congenital lipodystrophy 2 homolog; Bernardinelli-Seip congenital lipodystrophy type 2 protein; Bernardinelli-Seip congenital lipodystrophy type 2 protein homolog; bscl2; bscl2 {ECO:0000250; BSCL2, seipin lipid droplet biogenesis associated; G protein gamma 3 linked; Gng3lg; HMN5; MNCb-2630; PELD; seipin; SPG17; UniProtKB:Q96G97} | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 26580 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q96G97 | |
| BSCL2 | |
| E. coli-derived recombinant human Seipin/BSCL2 Asn279-Ser398. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction