missing translation for 'onlineSavingsMsg'
Learn More

Seipin/BSCL2 Antibody [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30734024 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734024 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734024 Supplier Novus Biologicals Supplier No. AF7385MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Seipin/BSCL2 Polyclonal antibody specifically detects Seipin/BSCL2 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Seipin/BSCL2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias Berardinelli-Seip congenital lipodystrophy 2 (seipin), Bernardinelli-Seip congenital lipodystrophy 2 (seipin), Bernardinelli-Seip congenital lipodystrophy type 2 protein, FLJ16651, GNG3LG, HMN5, MGC4694, seipin, spastic paraplegia 17 (Silver syndrome), SPG17
Host Species Sheep
Immunogen E. coli-derived recombinant human Seipin/BSCL2, Asn279-Ser398, Accession # Q96G97
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26580
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.