missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Segmentation protein paired Rabbit anti-Drosophila, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Segmentation protein paired Polyclonal specifically detects Segmentation protein paired in Drosophila samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Segmentation protein paired |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | PRD |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide corresponding to a region of Fruit fly (NP_723721). Peptide sequence MTVTAFAAAMHRPFFNGYSTMQDMNSGQGRVNQLGGVFINGRPLPNNIRL |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?