Learn More
Witeg™ Security Washing Bottle
Description
- With screw-threads
- Porosity of P0
- With two SVS-tubing connections, GL 14, center neck GL 32
- With septum
- Made of borosilicate glass
- Washing intensity of 130 L/h
Specifications
Specifications
| Capacity (Metric) | 500 mL |