missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ SecureFit™ X5000 Safety Helmet, Vented, Reflective, CE
Description
- Secure, comfortable head protection for working at heights and other hazardous industrial jobs
- Suspension system incorporates exclusive patented 3M™ Pressure Diffusion Technology
- 4-point chin strap selector system unique to 3M allows wearer to switch between EN 12492 (helmets for mountaineers) and EN 397 (industrial safety helmets) standards
- 6-point ratchet suspension allows wearer to customise height and fit with multiple levels of vertical adjustment
- Magnetic buckle clip as standard
- Inspired by climbing helmets with a brimless design to give you a clear upward view
- Accessory slots and clips are compatible with a variety of 3M face shields, earmuffs and other accessories
- 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV-radiation and it's time to replace it
- Certified to either EN12492 or EN397 (depending on chin strap configuration)
- Approved for lateral deformation (LD)
- UV-stabilized ABS
Specifications
Specifications
| Color | Yellow |
| For Use With (Application) | Construction, Forestry, Oil, and Gas |
| Material | ABS |
| Product Type | Safety Helmet |
| Suspension Type | Pressure Diffusion 6 Point Ratchet |
| Ventilated | Ventilated |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?