Learn More
3M™ SecureFit™ 100 Safety Glasses
Product Code. 17354690
Change view
Click to view available options
Model:
SF101AF-BLK-EU
SF101AF-BLU-EU
SF101AS-BLK-EU
SF101AS-BLU-EU
SF102AF-BLU-EU
SF103AF-BLU-EU
SF107AF-BLU-EU
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Unit Size:
Each
Pack of 20
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Lens Coating unitSize
17354690 SF101AF-BLK-EU Anti-fog, Scratch-resistant Pack of 20
17324680 SF103AF-BLU-EU Anti-fog, Scratch-resistant Each
17324690 SF101AS-BLK-EU Scratch-resistant Pack of 20
17334690 SF101AS-BLU-EU Scratch-resistant Each
17344690 SF102AF-BLU-EU Anti-fog, Scratch-resistant Each
17364690 SF107AF-BLU-EU Anti-fog, Scratch-resistant Each
17384690 SF101AF-BLU-EU Anti-fog, Scratch-resistant Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17354690 Supplier 3M™ Supplier No. 7100244060
Only null left
Add to basket
Edge
Add to basket
3M™ SecureFit™ 100 Series features a sleek, sporty lens shape coupled with a patented temple design that self-adjusts to different head sizes to provide a stylish yet comfortable fit for an extended period of time.
  • Economical robust 3M™ SecureFit™ Safety Glasses range
  • 3M's innovative SecureFit™ Pressure Diffusion Temple technology design features slotted ribs that provide comfortable pressure equalization. The temples flex and self-adjust to the size of wearer's head
  • Slim wrap around design to look better on smaller faces
  • Economical Bulk package (5-pack) available (black frame only)
  • Flat and flexible temple design combines well with 3M™ Protective Earmuffs
  • Wraparound design improves the security of fit and prevents slipping
  • Easy to clean
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance (Europe) EN 166:2001
Lens Tint Clear
Product Type Protective Eyewear
Frame Color Black
No. per Case 20
Model SF101AF-BLK-EU
Frame Material Polycarbonate
UV Protection UV400