Learn More
3M™ Secure Click™ Particulate Filters
Product Code. 16466390
Change view
Click to view available options
Filter Type:
P2R
P3R
Unit Size:
10 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filter Type unitSize
16466390 P2R 10 pairs
16426390 P3R 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16466390 Supplier 3M™ Supplier No. 7100197481
Only null left
Add to basket
Add to basket
3M™ Secure Click™ Particulate Filter is our CE approved filter for environments containing certain particles.
  • New dual flow cartridges and filters allow for easier breathing
  • Protects against particulates with optional nuisance level organic vapour/acid gas relief
  • Quick, simple connection. Simply align the connections and push until you hear a click
  • Use with 3M™ Secure Click™ Reusable Respirators 800 Series
  • Swept-back design allows an enhanced field of view and comfort
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved
Connection Type Secure Click™
Filter Type P2R
Product Type Filter
Model D3125
Quantity 80/Cs.