Learn More
3M™ Secure Click™ Filter Retainer D701
Product Code. 16496370
Change view
Click to view available options
:
For Use With: Reusable Respirators
Product Code.
16496370 For Use With: Reusable Respirators
1 options
Product Code. 16496370 Supplier 3M™ Supplier No. 7100184498
Only null left
Add to basket
Add to basket
3M™ Secure Click™ Filter Retainer D701 is designed for use with 3M™ Secure Click™ Respirators 800 Series.
  • Used to hold 3M™ Secure Click™ Particulate Filters D7N11 or D7P71 in place on 3M™ Secure Click™ Cartridges D8000 Series
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 3M™ Secure Click™ HF-800 Series Reusable Respirators
Quantity 100/Cs.