missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Secukinumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
£397.00 - £1002.00
Specifications
| Antigen | Secukinumab |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284558
|
Invitrogen™
MA559017 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30282986
|
Invitrogen™
MA559018 |
1 mg |
£1002.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Secukinumab (Cosentyx) is a human monoclonal antibody designed for the treatment of uveitis, rheumatoid arthritis, ankylosing spondylitis, and psoriasis. Secukinumab is an interleukin-17A (IL-17A) inhibitor marketed by Novartis. IL-17 is a group of proinflammatory cytokines released by cells of the immune system and exist in higher levels in many immune conditions associated with chronic inflammation. By targeting IL-17A, secukinumab has shown excellent efficacy in psoriasis by normalizing skin histology and was approved by the United States Food and pharmaceutical Administration on January 21, 2015 to treat adults with moderate-to-severe plaque psoriasis.Specifications
| Secukinumab | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG1 | |
| Human | |
| AIN457; Cosentyx | |
| Antibody |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM Histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title