missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SECTM1 Antibody (106), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90085R
This item is not returnable.
View return policy
Description
SECTM1 Monoclonal specifically detects SECTM1 in Human samples. It is validated for ELISA.
Specifications
| SECTM1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 106 | |
| ELISA | |
| K12secreted and transmembrane protein 1, Protein K-12, secreted and transmembrane 1, type 1a transmembrane protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human SECTM1 (Q8WVN6-1; Met1-Gly145). | |
| 0.1 mL | |
| Cytokine Research | |
| 6398 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction