missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Section Lifter

Product Code. 10075030
Change view
Click to view available options
Length (Metric):
150 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10075030 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10075030 Supplier Bochem™ Supplier No. 3532
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Thin-bladed, flexible section lifter. Bochem™ Section Lifter is constructed of 18/10 stainless steel and features an ergonomic wooden handle.

  • Thin blade.
  • 18/10 stainless steel construction.
  • Ergonomic wooden handle.

Specifications

Product Type Section Lifter
Length (Metric) Blade 50 mm
Width (Metric) Blade 10 mm
Handle Material Wood
Material Stainless Steel (18/10)
Length (Metric) 150 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.