missing translation for 'onlineSavingsMsg'
Learn More

Secretogranin V Antibody (8G11), Novus Biologicals™

Product Code. 18420300 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420300 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420300 Supplier Novus Biologicals Supplier No. H00006447M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Secretogranin V Monoclonal antibody specifically detects Secretogranin V in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Secretogranin V
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 8G11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003011
Gene Alias neuroendocrine protein 1 (7B2 protein), Pituitary polypeptide, prohormone convertase chaperone, Secretogranin V, secretogranin V (7B2 protein), Secretogranin-5, Secretory granule endocrine protein I
Host Species Mouse
Immunogen SCG5 (NP_003011, 59 a.a. ~ 159 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EYPAHQAMNLVGPQSIEGGAHEGLQHLGPFGNIPNIVAELTGDNIPKDFSEDQGYPDPPNPCPVGKTDDGCLENTPDTAEFSREFQLHQHLFDPEHDYPGL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6447
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.