missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Secretin receptor Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14172-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a G protein-coupled receptor and belongs to the glucagon-VIP-secretin receptor family. It binds secretin which is the most potent regulator of pancreatic bicarbonate, electrolyte and volume secretion. Secretin and its receptor are suggested to be involved in pancreatic cancer and autism.Specifications
| Secretin receptor | |
| Polyclonal | |
| Unconjugated | |
| SCTR | |
| SCT R, SCTR, Secretin receptor, SR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 319229, 6344, 81779 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P23811, P47872, Q5FWI2 | |
| Sctr | |
| Secretin receptor Fusion Protein Ag5371 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction