missing translation for 'onlineSavingsMsg'
Learn More

Secoisolariciresinol diglucoside, MedChemExpress

Product Code. 16496717
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496717 5 mg 5mg
16475006 100 mg 100mg
16474008 10 mg 10mg
16432936 25 mg 25mg
16420367 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496717 Supplier Medchem Express Supplier No. HY105008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Secoisolariciresinol diglucoside ((S,S)-SDG), the main lignan in wholegrain flaxseed, is known for its beneficial effects including anti-inflammatory, antioxidant, anti-mutagenic, anti-microbial, anti-obesity, hypolipidemic, and neuroprotective effects.

Secoisolariciresinol diglucoside ((S,S)-SDG), the main lignan in wholegrain flaxseed, is known for its beneficial effects including anti-inflammatory, antioxidant, anti-mutagenic, anti-microbial, anti-obesity, hypolipidemic, and neuroprotective effects.

Specifications

Chemical Name or Material Secoisolariciresinol diglucoside
CAS 257930-74-8
Color White
Purity Grade Notes Research
Molecular Formula C32H46O16
Quantity 5 mg
Synonym (S,S)-SDG (S,S)-LGM2605
Solubility Information H2O : ≥ 100 mg/mL (145.62 mM)
SMILES COC1=C(O)C=CC(C[C@H](CO[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)CO)[C@](CO[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)CO)([H])CC4=CC(OC)=C(O)C=C4)=C1
Molecular Weight (g/mol) 686.7
Formula Weight 686.7
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.