missing translation for 'onlineSavingsMsg'
Learn More

seco-(8,9)-Buspirone Carboxylic Acid, TRC

Product Code. 30137974
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137974 1 g 1g
30132999 250 mg 250mg
30005912 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137974 Supplier Toronto Research Chemicals Supplier No. TRCB6897151G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 257877-43-3
Molecular Formula C21 H33 N5 O3
Molecular Weight (g/mol) 403.52
Synonym 1-[2-Oxo-2-[[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]amino]ethyl]cyclopentaneacetic acid
IUPAC Name 2-[1-[2-oxo-2-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butylamino]ethyl]cyclopentyl]acetic acid
SMILES OC(=O)CC1(CC(=O)NCCCCN2CCN(CC2)c3ncccn3)CCCC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H33N5O3/c27-18(16-21(17-19(28)29)6-1-2-7-21)22-8-3-4-11-25-12-14-26(15-13-25)20-23-9-5-10-24-20/h5,9-10H,1-4,6-8,11-17H2,(H,22,27)(H,28,29)
Formula Weight 403.2583
Chemical Name or Material seco-(8,9)-Buspirone Carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.