missing translation for 'onlineSavingsMsg'
Learn More

Secnidazole, Thermo Scientific™

Product Code. p-7227423 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17780639 1 g 1g
17790639 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17780639 Supplier Thermo Scientific Acros Supplier No. 469030010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material Secnidazole
Type Secnidazole
Physical Form Powder
CAS 3366-95-8
Melting Point 70°C to 76°C
Infrared Spectrum Conforms
Packaging Glass Bottle
Water ≤6 %
Color White to Yellow
Quantity 1 g
Molecular Formula C7 H11 N3 O3
Product Line Life science
MDL Number 00864656
Shipping Condition No special requirements
Merck Index XIII 8494
Solubility Information Soluble in methanol
Molecular Weight (g/mol) 185.18
Percent Purity ≥97.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.