missing translation for 'onlineSavingsMsg'
Learn More

Seclidemstat, MedChemExpress

Product Code. 16484936
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16484936

Brand: Medchem Express HY103713

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Seclidemstat is a potent noncompetitive and reversible KDM1A (LSD1) inhibitor (Ki=31 nM, IC50=13 nM). Seclidemstat promotes antitumor immunity in switch/sucrose nonfermentable (SWI/SNF) complex mutated ovarian cancer, as well as inhibit virus production, viral DNA replication, and late gene expression. Seclidemstat can be used for the research of Ewing Sarcoma.

Seclidemstat is a potent noncompetitive and reversible KDM1A (LSD1) inhibitor (Ki=31 nM, IC50=13 nM). Seclidemstat promotes antitumor immunity in switch/sucrose nonfermentable (SWI/SNF) complex mutated ovarian cancer, as well as inhibit virus production, viral DNA replication, and late gene expression. Seclidemstat can be used for the research of Ewing Sarcoma.

Specifications

Chemical Name or Material Seclidemstat
CAS 1423715-37-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H23ClN4O4S
Quantity 100 mg
Synonym SP-2577
Solubility Information DMSO : 50 mg/mL (110.88 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(N/N=C(C1=CC(Cl)=CC=C1O)\C)C2=CC=CC(S(=O)(N3CCN(C)CC3)=O)=C2
Molecular Weight (g/mol) 450.94
Formula Weight 450.94
Percent Purity 98.22%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.