missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SECISBP2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-53795
This item is not returnable.
View return policy
Description
SECISBP2 Polyclonal specifically detects SECISBP2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Peptide ELISA, Knockdown Validated.
Specifications
| SECISBP2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| DKFZp686C09169, SBP2SECIS-binding protein 2, SECIS binding protein 2, selenocysteine insertion sequence binding protein 2, selenocysteine insertion sequence-binding protein 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 79048 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ELISA, KnockDown | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/mL, Immunohistochemistry 3 ug/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 3 ug/mL, Peptide ELISA Detection limit 1:64000, Knockdown Validated | |
| NP_076982.3 | |
| SECISBP2 | |
| Peptide with sequence C-KERQERKQRLQEN corresponding to internal region according to NP_076982.3. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Dog, Pig and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction