missing translation for 'onlineSavingsMsg'
Learn More

SECISBP2 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18422438 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422438 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422438 Supplier Novus Biologicals Supplier No. NBP222301AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SECISBP2 Polyclonal antibody specifically detects SECISBP2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SECISBP2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp686C09169, SBP2SECIS-binding protein 2, SECIS binding protein 2, selenocysteine insertion sequence binding protein 2, selenocysteine insertion sequence-binding protein 2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 804 to 854 of human SECIS Binding Protein 2 using the numbering given in entry NP_076982.3 (GeneID 79048).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79048
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.