missing translation for 'onlineSavingsMsg'
Learn More

Secernin-1 Antibody (OTI1F2), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18400187 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400187 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400187 Supplier Novus Biologicals Supplier No. NBP274052AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Secernin-1 Monoclonal antibody specifically detects Secernin-1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Secernin-1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1F2
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias KIAA0193SES1, secernin 1, secernin-1
Host Species Mouse
Immunogen Full length recombinant protein of human Secernin-1 (NP_055581) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9805
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.