missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Secernin-1 Antibody (OTI1F2), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74052AF532
This item is not returnable.
View return policy
Description
Secernin-1 Monoclonal antibody specifically detects Secernin-1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Secernin-1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| KIAA0193SES1, secernin 1, secernin-1 | |
| Full length recombinant protein of human Secernin-1 (NP_055581) produced in HEK293T cells. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 9805 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1F2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction