missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Secernin-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77187-0.025mg
This item is not returnable.
View return policy
Description
Secernin-1 Polyclonal specifically detects Secernin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Secernin-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| KIAA0193SES1, secernin 1, secernin-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9805 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| NP_001138985 | |
| SCRN1 | |
| Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human SCRN1. The immunogen is located within amino acids 320 - 370 of SCRN1. Amino Acid Squence: CPAKKEPRFQEKPDRRHELYK | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction