missing translation for 'onlineSavingsMsg'
Learn More

Secalciferol, MedChemExpress

Product Code. 16481919
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481919 1 mg 1mg
16450498 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481919 Supplier Medchem Express Supplier No. HY32343
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Secalciferol is a metabolite of Vitamin D, a possibly anti-inflammatory steroid which is involved in bone ossification.

Secalciferol is a metabolite of Vitamin D, a possibly anti-inflammatory steroid which is involved in bone ossification.

Specifications

Chemical Name or Material Secalciferol
CAS 55721-11-4
Color White
Purity Grade Notes Research
Molecular Formula C27H44O3
Quantity 1 mg
Synonym (24R)-24,25-Dihydroxyvitamin D3
Solubility Information DMSO : 100 mg/mL (240.02 mM; Need ultrasonic)
SMILES C=C(CC1)/C(C[C@H]1O)=C\C=C2[C@@]3([H])[C@@](CCC\2)(C)[C@@H]([C@H](C)CC[C@@H](O)C(C)(C)O)CC3
Molecular Weight (g/mol) 416.64
Formula Weight 416.64
Percent Purity 98.58%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.