missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sec8 Antibody (14G1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
£138.00 - £558.00
Specifications
| Antigen | Sec8 |
|---|---|
| Clone | 14G1 |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 12.5 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18078139
|
Novus Biologicals
NBP1-97492-0.025MG |
0.025 mg |
£138.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18097218
|
Novus Biologicals
NBP1-97492-0.05MG |
0.05 mg |
£296.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18121010
|
Novus Biologicals
NBP1-97492-0.2MG |
0.2 mg |
£558.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sec8 Monoclonal specifically detects Sec8 in Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Sec8 | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 12.5 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| 60412 | |
| Recombinant rat rSec8. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 14G1 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Bovine, Canine, Fish, Hamster, Primate, Guinea Pig, Rabbit, Sheep, Mouse, Human, Rat, Chicken | |
| exocyst complex component 4, KIAA1699SEC8L1, MGC27170, REC8, SEC8Exocyst complex component Sec8, SEC8-like 1, SEC8-like 1 (S. cerevisiae), Sec8p | |
| EXOC4 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title