missing translation for 'onlineSavingsMsg'
Learn More

SEC62 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18403525 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403525 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403525 Supplier Novus Biologicals Supplier No. NBP281813JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SEC62 Polyclonal antibody specifically detects SEC62 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEC62
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Dtrp1, hTP-1, S.cerevisiae, homolog of, SEC62 homolog (S. cerevisiae), TLOC1, TP-1, translocation protein 1, translocation protein SEC62
Host Species Rabbit
Immunogen SEC62 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SEC62. The immunogen is located within amino acids 270 - 320 of SEC62. Amino Acid Squence: TYTHEYKGPKADLKKDEKS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7095
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.